4-Fluoroindolin-2-one 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Fluoro-2-oxindole, 4-Fluorooxindole

PC8677-10G PC8677-1G PC8677-5G
76609-512EA 74.38 USD
76609-512 76609-514 76609-516
4-Fluoroindolin-2-one 97%
4-Fluoroindolin-2-one
Formula: C₈H₆FNO
Storage Temperature: Ambient
MDL Number: MFCD04112528
CAS Number: 138343-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR