Chlorodimethyl(pentafluorophenyl)silane (flophemesyl chloride) 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC5664-25G PC5664-5G
76618-786EA 100.84 USD
76618-786 76618-788
Chlorodimethyl(pentafluorophenyl)silane (flophemesyl chloride) 98%
Chlorodimethyl(pentafluorophenyl)silane (Flophemesyl chloride)
Formula: C₈H₆ClF₅Si
Boiling Pt: 88…90 °C/10mmHg
Density: 1.381 g/cm³ (25 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00000500
CAS Number: 20082-71-7
EINECS: 243-506-9
UN: 2987
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR