4-(Trifluoromethoxy)benzamidoxime 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: N-Hydroxy-4-(trifluoromethoxy)benzimidamide

PC7436D-1G PC7436D-25G PC7436D-5G
76608-516EA 20.26 USD
76608-516 76608-518 76608-520
4-(Trifluoromethoxy)benzamidoxime 97%
4-(Trifluoromethoxy)benzamidoxime
Formula: C₈H₇F₃N₂O₂
Melting Pt: 113…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00085175
CAS Number: 56935-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR