Tris(perfluorobutyl)amine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Perfluorotri-n-butylamine, Heptacosafluorotri-n-butylamine, Perfluorotributylamine, FC-43, Heptacosafluorotributylamine, PFTBA, Tris(nonafluorobutyl)amine

PC6220-100G PC6220-25G
76603-566EA 82.52 USD
76603-566 76603-568
Tris(perfluorobutyl)amine
Tris(perfluorobutyl)amine
Formula: C₁₂F₂₇N
MW: 671.1
Boiling Pt: 155
Melting Pt: -55
Density: 1.9 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000436
CAS Number: 311-89-7
EINECS: 206-223-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR