2-Fluoro-6-nitrobenzoic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC5538-100G PC5538-25G PC5538-5G
76602-080EA 93.04 USD
76602-080 76602-082 76602-084
2-Fluoro-6-nitrobenzoic acid 97%
2-Fluoro-6-nitrobenzoic acid
Formula: C₇H₄FNO₄
Melting Pt: 143…147 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01862079
CAS Number: 385-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR