2-Amino-6-(trifluoromethyl)pyridine 97+%

Supplier: APOLLO SCIENTIFIC
Danger

PC9615-100G PC9615-25G PC9615-5G
76619-042EA 94.74 USD
76619-042 76619-044 76619-046
2-Amino-6-(trifluoromethyl)pyridine 97+%
2-Amino-6-(trifluoromethyl)pyridine
Formula: C₆H₅F₃N₂
Melting Pt: 85…89 °C
Storage Temperature: Ambient
MDL Number: MFCD03093868
CAS Number: 34486-24-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR