(Trifluoromethyl)trimethylsilane 99%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC7779-100G
PC7779-25G
PC7779-5G
76614-596EA
96.78
USD
76614-596
76614-598
76614-600
(Trifluoromethyl)trimethylsilane 99%
(Trifluoromethyl)trimethylsilane
Ruppert's reagent - valuable trifluoromethylating agent of electrophilic compounds.
Formula:
C₄H₉F₃Si Boiling Pt: 54…55 °C Density: 0.962 g/cm³ (25 °C) Flash Pt: –17 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00145454 CAS Number: 81290-20-2 UN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...