1-Fluoro-4-ethynylbenzene 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Ethynyl-4-fluorobenzene, (4-Fluorophenyl)acetylene, (4-Fluorophenyl)ethyne

PC5955-1G PC5955-25G PC5955-5G
76614-372EA 20.28 USD
76614-372 76614-374 76614-376
1-Fluoro-4-ethynylbenzene 98%
1-Fluoro-4-ethynylbenzene
Formula: C₈H₅F
Boiling Pt: 55…56 °C/40mmHg
Melting Pt: 27…28 °C
Density: 1.048 g/cm³ (25 °C)
Flash Pt: 29 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00168823
CAS Number: 766-98-3
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR