Triethylsilyl trifluoromethanesulfonate 99%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
Triethylsilyl trifluoromethanesulfonate, Triethylsilyl trifluoromethanesulphonate
PC7114-100G
PC7114-25G
PC7114-5G
76627-374EA
58.06
USD
76627-374
76627-376
76627-378
Triethylsilyl trifluoromethanesulfonate 99%
Triethylsilyl trifluoromethanesulphonate
Lewis acid. Powerful silylating reagent .
Formula:
C₇H₁₅F₃O₃SSi MW: 264.34 g/mol Boiling Pt: 207 °C Density: 1.169 g/cm³ (25 °C) Flash Pt: 66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000407 CAS Number: 79271-56-0 EINECS: 279-124-4 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...