4-(Trifluoromethyl)thiophenol 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Mercaptobenzotrifluoride, 4-(Trifluoromethyl)benzene-1-thiol, 4-(Trifluoromethyl)benzenethiol

PC7773-25G PC7773-50G PC7773-5G
76608-820EA 391.16 USD
76608-820 76608-822 76608-824
4-(Trifluoromethyl)thiophenol 97%
4-(Trifluoromethyl)thiophenol
Formula: C₇H₅F₃S
Boiling Pt: 66…68 °C/15mmHg
Density: 1.3 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00042320
CAS Number: 825-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR