2-Amino-4-fluorophenol 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Fluoro-2-hydroxyaniline

PC5940-1G PC5940-5G
76603-368EA 20.87 USD
76603-368 76603-370
2-Amino-4-fluorophenol 98%
2-Amino-4-fluorophenol
Formula: C₆H₆FNO
Melting Pt: 138…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00077451
CAS Number: 399-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR