Xenon difluoride 99,9%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Xenon(II) fluoride

PC8020-25G PC8020-5G
76618-942EA 74 USD
76618-942 76618-944 76618-946
Xenon difluoride 99,9%
Xenon difluoride

Convenient fluorinating agent

Formula: F₂Xe
Melting Pt: 129 °C
Density: 4.32 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00040538
CAS Number: 13709-36-9
EINECS: 237-251-2
UN: 3087
ADR: 5.1+6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR