2,3,6-Trifluoroaniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC7258L-5G PC7258L-1G PC7258L-25G
76608-288EA 107.98 USD
76608-288 76608-284 76608-286
2,3,6-Trifluoroaniline 98%
2,3,6-Trifluoroaniline
Formula: C₆H₄F₃N
MW: 147.1
Boiling Pt: 150…151 °C
Density: 1.39 g/cm³ (25 °C)
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00012369
CAS Number: 67815-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR