2,4,5-Trifluorobenzonitrile 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC7283M-100G PC7283M-25G
76627-398EA 169.96 USD
76627-398 76627-400 76627-402
2,4,5-Trifluorobenzonitrile 99%
2,4,5-Trifluorobenzonitrile
Formula: C₇H₂F₃N
Boiling Pt: 168…170 °C
Density: 1.373 g/cm³ (25 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00013289
CAS Number: 98349-22-5
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR