6-Fluoro-3-methyl-2-(trifluoromethyl)benzoic acid 97%

Supplier: APOLLO SCIENTIFIC

PC303397-1G PC303397-5G
76599-770EA 58.06 USD
76599-770 76599-772
6-Fluoro-3-methyl-2-(trifluoromethyl)benzoic acid 97%
6-Fluoro-3-methyl-2-(trifluoromethyl)benzoic acid
Formula: C₉H₆F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD32640056
CAS Number: 2383927-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR