4-Chloro-2-fluoro-5-methylbenzoic acid 97%

Supplier: APOLLO SCIENTIFIC

PC303325-1G PC303325-250MG
76599-658EA 282.18 USD
76599-658 76599-660
4-Chloro-2-fluoro-5-methylbenzoic acid 97%
4-Chloro-2-fluoro-5-methylbenzoic acid
Formula: C₈H₆ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD20660427
CAS Number: 177211-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR