2-Chloro-6-(trifluoromethyl)benzenesulfonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC303244-1G PC303244-5G
76613-598EA 254.68 USD
76613-598 76613-600
2-Chloro-6-(trifluoromethyl)benzenesulfonyl chloride 97%
2-Chloro-6-(trifluoromethyl)benzenesulfonyl chloride
Formula: C₇H₃Cl₂F₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD06408806
CAS Number: 874814-70-7
UN: 1759
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR