5-Fluoro-2-(trifluoromethyl)benzenesulfonyl chloride 97%

Supplier: APOLLO SCIENTIFIC

PC303210-1G PC303210-250MG
76613-574EA 681.48 USD
76613-574 76613-576
5-Fluoro-2-(trifluoromethyl)benzenesulfonyl chloride 97%
5-Fluoro-2-(trifluoromethyl)benzenesulfonyl chloride
Formula: C₇H₃ClF₄O₂S
Storage Temperature: Ambient
MDL Number: MFCD13185646
CAS Number: 915763-87-0
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR