2-Chloro-5-methoxy-3-(trifluoromethyl)aniline 97%

Supplier: APOLLO SCIENTIFIC

PC303191-1G PC303191-250MG
76613-558EA 660.72 USD
76613-558 76613-560
2-Chloro-5-methoxy-3-(trifluoromethyl)aniline 97%
2-Chloro-5-methoxy-3-(trifluoromethyl)aniline
Formula: C₈H₇ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD27986849
CAS Number: 1706436-22-7
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR