4-Methoxy-3-methyl-5-(trifluoromethyl)phenylacetic acid 97%

Supplier: APOLLO SCIENTIFIC

PC303138-1G PC303138-250MG
76599-398EA 635.51 USD
76599-398 76599-400
4-Methoxy-3-methyl-5-(trifluoromethyl)phenylacetic acid 97%
4-Methoxy-3-methyl-5-(trifluoromethyl)phenylacetic acid
Formula: C₁₁H₁₁F₃O₃
MW: 248.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27986835
CAS Number: 1706446-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR