3-(2,4,5-Trifluorophenyl)propionic acid 98%

Supplier: APOLLO SCIENTIFIC

PC302937-10G PC302937-1G PC302937-5G
76599-016EA 292.36 USD
76599-016 76599-018 76599-020
3-(2,4,5-Trifluorophenyl)propionic acid 98%
3-(2,4,5-Trifluorophenyl)propionic acid
Formula: C₉H₇F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD06660217
CAS Number: 651047-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR