4-Chloro-3-(trifluoromethyl)benzamide 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC302410-1G PC302410-5G PC302410-10G
76597-668EA 20.77 USD
76597-668 76597-670 76597-666
4-Chloro-3-(trifluoromethyl)benzamide 97%
4-Chloro-3-(trifluoromethyl)benzamide
Formula: C₈H₅ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD04115832
CAS Number: 62584-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR