5-Chloro-2-methyl-4-(trifluoromethoxy)benzenesulfonyl chloride 97%

Supplier: APOLLO SCIENTIFIC

PC302409-1G PC302409-250MG
76613-232EA 1760.26 USD
76613-232 76613-234
5-Chloro-2-methyl-4-(trifluoromethoxy)benzenesulfonyl chloride 97%
5-Chloro-2-methyl-4-(trifluoromethoxy)benzenesulfonyl chloride
Formula: C₈H₅Cl₂F₃O₃S
Storage Temperature: Ambient
MDL Number: MFCD28054251
CAS Number: 1706447-06-4
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR