4-Chloro-2-(trifluoromethyl)benzyl alcohol 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC302408-10G PC302408-1G PC302408-5G
76597-660EA 261.8 USD
76597-660 76597-662 76597-664
4-Chloro-2-(trifluoromethyl)benzyl alcohol 98%
4-Chloro-2-(trifluoromethyl)benzyl alcohol
Formula: C₈H₆ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD04115839
CAS Number: 773872-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR