2-Chloro-3,6-difluoro-DL-phenylalanine

Supplier: APOLLO SCIENTIFIC

PC302125-1G PC302125-5G
76596-890EA 110.02 USD
76596-890 76596-892
2-Chloro-3,6-difluoro-DL-phenylalanine
2-Chloro-3,6-difluoro-DL-phenylalanine
Formula: C₉H₈ClF₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD04116007
CAS Number: 1259981-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR