3-Methoxy-5-(trifluoromethyl)phenol 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: 3-hydroxy-5-methoxy-benzotrifluoride

PC302309-1G PC302309-25G PC302309-5G
76597-412EA 41.76 USD
76597-412 76597-414 76597-416
3-Methoxy-5-(trifluoromethyl)phenol 98%
3-Methoxy-5-(trifluoromethyl)phenol
Formula: C₈H₇F₃O₂
Melting Pt: 74…76 °C
Storage Temperature: Ambient
MDL Number: MFCD09025418
CAS Number: 349-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR