4-Methoxy-3-methyl-5-(trifluoromethyl)aniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC303129-1G PC303129-5G
76613-502EA 101.88 USD
76613-502 76613-504
4-Methoxy-3-methyl-5-(trifluoromethyl)aniline 98%
4-Methoxy-3-methyl-5-(trifluoromethyl)aniline
Formula: C₉H₁₀F₃NO
Storage Temperature: Ambient
MDL Number: MFCD22682815
CAS Number: 1431329-84-8
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR