4-Fluoro-3-methyl-5-(trifluoromethyl)benzamide 97%

Supplier: APOLLO SCIENTIFIC

Synonyms: 4-Fluoro-3-methyl-5-(trifluoromethyl)benzamide

PC302880-1G PC302880-5G
76598-856EA 82.52 USD
76598-856 76598-858
4-Fluoro-3-methyl-5-(trifluoromethyl)benzamide 97%
4-Fluoro-3-methyl-5-(trifluoromethyl)benzamide
Formula: C₉H₆F₄NO
Storage Temperature: Ambient
MDL Number: MFCD22201066
CAS Number: 1373920-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR