2-Methyl-5-(trifluoromethyl)benzaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC302820-5G PC302820-1G
76598-748EA 124.28 USD
76598-748 76598-746
2-Methyl-5-(trifluoromethyl)benzaldehyde 98%
2-Methyl-5-(trifluoromethyl)benzaldehyde
Formula: C₉H₇F₃O
Storage Temperature: Ambient
MDL Number: MFCD06660200
CAS Number: 886498-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR