2-Bromo-4-chloro-5-fluoroaniline 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC303362-5G
PC303362-1G
76599-694EA
86
USD
76599-694
76599-696
2-Bromo-4-chloro-5-fluoroaniline 98%
2-Bromo-4-chloro-5-fluoroaniline
Formula:
C₆H₄BrClFN MW: 224.458 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12026231 CAS Number: 120694-11-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...