3-Fluoro-5-methylbenzylamine 98%

Supplier: APOLLO SCIENTIFIC

PC302093-10G PC302093-1G PC302093-5G
76613-148EA 542.94 USD
76613-148 76613-150 76613-152
3-Fluoro-5-methylbenzylamine 98%
3-Fluoro-5-methylbenzylamine
Formula: C₈H₁₀FN
Storage Temperature: Ambient
MDL Number: MFCD06213636
CAS Number: 771573-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR