1,4-Diiodotetrafluorobenzene 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
1,2,4,5-Tetrafluoro-3,6-diiodobenzene
PC2970-1G
PC2970-25G
PC2970-5G
76595-972EA
26
USD
76595-972
76595-974
76595-976
1,4-Diiodotetrafluorobenzene 97%
1,4-Diiodotetrafluorobenzene
Formula:
C₆F₄I₂ MW: 401.87 g/mol Melting Pt: 107…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001055 CAS Number: 392-57-4 EINECS: 206-877-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...