3-Bromo-2-chloro-6-fluorobenzyl fluoride 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC27904-1G PC27904-250MG
76622-536EA 578.6 USD
76622-536 76622-538
3-Bromo-2-chloro-6-fluorobenzyl fluoride 95%
3-Bromo-2-chloro-6-fluorobenzyl fluoride
Formula: C₇H₄BrClF₂
Storage Temperature: Ambient
MDL Number: MFCD31664896
CAS Number: 2244083-98-3
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR