3-Amino-4-methylbenzotrifluoride 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Methyl-5-(trifluoromethyl)aniline

PC2210-1G PC2210-25G PC2210-5G
76617-846EA 20.87 USD
76617-846 76617-848 76617-850
3-Amino-4-methylbenzotrifluoride 97%
3-Amino-4-methylbenzotrifluoride
Formula: C₈H₈F₃N
MW: 175.16 g/mol
Melting Pt: 36…40 °C
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00134580
CAS Number: 25449-96-1
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR