3-Chloro-4-fluorobenzotrifluoride 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Chloro-2-fluoro-5-(trifluoromethyl)benzene, 3-Chloro-4-fluorobenzotrifluoride, 2-Chloro-1-fluoro-4-(trifluoromethyl)benzene

PC1863L-100G PC1863L-25G PC1863L-500G
76612-922EA 90.68 USD
76612-922 76612-924 76612-926
3-Chloro-4-fluorobenzotrifluoride 98%
3-Chloro-4-fluorobenzotrifluoride
Formula: C₇H₃ClF₄
Boiling Pt: 137 °C
Density: 1.49 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00042207
CAS Number: 78068-85-6
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR