Barium fluoride 99%

Supplier: APOLLO SCIENTIFIC
Danger

PC1170-100G PC1170-250G
76612-778EA 57.04 USD
76612-778 76612-780
Barium fluoride 99%
Barium fluoride
Formula: BaF₂
Boiling Pt: 2260 °C
Melting Pt: 1353 °C
Density: 4.89 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003450
CAS Number: 7787-32-8
EINECS: 232-108-0
UN: 1564
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR