(2-Chloro-4-fluorophenyl)methanesulfonyl chloride

Supplier: APOLLO SCIENTIFIC

PC110117-1G PC110117-250MG
76622-384EA 166.04 USD
76622-384 76622-386
(2-Chloro-4-fluorophenyl)methanesulfonyl chloride
(2-Chloro-4-fluorophenyl)methanesulphonyl chloride
Formula: C₇H₅Cl₂FO₂S
Storage Temperature: Ambient
CAS Number: 541528-45-4
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR