2-Amino-5-chlorophenol 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Chloro-2-hydroxyaniline

OR9877-100G OR9877-25G OR9877-5G
76590-872EA 119.18 USD
76590-872 76590-870 76590-868
2-Amino-5-chlorophenol 97%
2-Amino-5-chlorophenol
Formula: C₆H₆ClNO
MW: 143.57 g/mol
Melting Pt: 145…155 °C
Storage Temperature: Ambient
MDL Number: MFCD02093863
CAS Number: 28443-50-7
EINECS: 249-020-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR