2-Amino-1-phenylethanol, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: alpha-(Aminomethyl)benzyl Alcohol, DL-β-Hydroxyphenethylamine, Phenylethanolamine

OR9824-100G OR9824-25G OR9824-500G
76617-522EA 75.4 USD
76617-522 76617-524 76617-526
2-Amino-1-phenylethanol, Technical Grade
2-Amino-1-phenylethanol
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 160 °C/17mmHg
Melting Pt: 56…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00008137
CAS Number: 7568-93-6
EINECS: 231-469-1
UN: 3259
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR