2-Amino-6-bromophenol 97+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Bromo-2-hydroxyaniline

OR9740-100G OR9740-25G OR9740-5G
76590-760EA 627.5 USD
76590-760 76590-758 76590-756
2-Amino-6-bromophenol 97+%
2-Amino-6-bromophenol
Formula: C₆H₆BrNO
MW: 188.02 g/mol
Melting Pt: 83…85 °C
Storage Temperature: Ambient
MDL Number: MFCD08277300
CAS Number: 28165-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR