3-Amino-2,6-dimethylpyridine 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,6-Dimethylpyridin-3-amine

OR9514-100G OR9514-25G OR9514-5G
76617-486EA 154.84 USD
76617-486 76617-488 76617-490
3-Amino-2,6-dimethylpyridine 98%
3-Amino-2,6-dimethylpyridine
Formula: C₇H₁₀N₂
Boiling Pt: 230 °C
Melting Pt: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD01646409
CAS Number: 3430-33-9
EINECS: 222-332-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR