3-Amino-1-adamantanol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Aminoadamantan-1-ol

OR9509-100G OR9509-25G
76590-352EA 25 USD
76590-352 76590-350 76590-348
3-Amino-1-adamantanol
3-Amino-1-adamantanol
Formula: C₁₀H₁₇NO
MW: 167.25 g/mol
Melting Pt: 263…269 °C (dec.)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01821204
CAS Number: 702-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR