5-Aminobenzo[b]furan 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Benzo[b]furan-5-amine, 1-Benzofuran-5-amine

OR9276-1G OR9276-250MG OR9276-5G
76589-882EA 168.08 USD
76589-882 76589-880 76589-884
5-Aminobenzo[b]furan 97%
5-Aminobenzo[b]furan
Formula: C₈H₇NO
Storage Temperature: Freezer
MDL Number: MFCD04125391
CAS Number: 58546-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR