3-Amino-2-bromophenol 95+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-amino-2-bromophenol

OR924467-1G OR924467-250MG
76589-802EA 50.94 USD
76589-802 76589-800
3-Amino-2-bromophenol 95+%
3-Amino-2-bromophenol
Formula: C₆H₆BrNO
MW: 188.022 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846453
CAS Number: 100367-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR