Triphenylphosphine 99+%

Supplier: APOLLO SCIENTIFIC
Warning

OR9088-1KG OR9088-250G OR9088-5KG
76589-496EA 74.38 USD
76589-496 76589-494 76589-498
Triphenylphosphine 99+%
Triphenylphosphine
Formula: C₁₈H₁₅P
MW: 262.29 g/mol
Boiling Pt: 377
Melting Pt: 79
Density: 1.2 g/cm³ (25 °C)
Flash Pt: 181 °C
Storage Temperature: Ambient
MDL Number: MFCD00003043
CAS Number: 603-35-0
EINECS: 210-036-0
Merck Index: 12,09873

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR