(S)-(-)-2-Pyrrolidinone-5-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR8965-100G OR8965-500G
76589-264EA 20.32 USD
76589-264 76589-266
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid
Formula: C₅H₇NO₃
Melting Pt: 155…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00005272
CAS Number: 98-79-3
EINECS: 202-700-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR