4-Amino-3-bromobenzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

OR7631-100G OR7631-25G
76625-452EA 103.9 USD
76625-452 76625-454
4-Amino-3-bromobenzonitrile
4-Amino-3-bromobenzonitrile
Formula: C₇H₅BrN₂
Melting Pt: 106…112 °C
Storage Temperature: Ambient
MDL Number: MFCD01861386
CAS Number: 50397-74-5
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR