2,6-Dimethylbenzylamine 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR70001-1G OR70001-5G
76612-370EA 38.72 USD
76612-370 76612-372
2,6-Dimethylbenzylamine 98%
2,6-Dimethylbenzylamine
Formula: C₉H₁₃N
Boiling Pt: 111…114 °C/15mmHg
Storage Temperature: Ambient
CAS Number: 74788-82-2
UN: 2735
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR