4-Amino-2,2,6,6-tetramethylpiperidine 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR6937-100G OR6937-500G
76612-360EA 58.06 USD
76612-360 76612-362 76612-364
4-Amino-2,2,6,6-tetramethylpiperidine 98%
4-Amino-2,2,6,6-tetramethylpiperidine
Formula: C₉H₂₀N₂
MW: 156.27 g/mol
Boiling Pt: 188…189 °C
Melting Pt: 16…18 °C
Density: 0.912 g/cm³ (25 °C)
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00005984
CAS Number: 36768-62-4
EINECS: 253-197-2
UN: 2735
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR