Methyl-4-(aminomethyl)benzoate hydrochloride
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
4-(Aminomethyl)benzoic Acid Methyl Ester Hydrochloride
, Methyl 4-(aminomethyl)benzoate hydrochloride
OR6787-5G
OR6787-25G
76595-144EA
22
USD
76595-144
76595-146
Methyl-4-(aminomethyl)benzoate hydrochloride
Methyl-4-(aminomethyl)benzoate hydrochloride
Formula:
C₉H₁₁NO₂ MW: 201.65 g/mol Melting Pt: 234…243 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00182671 CAS Number: 6232-11-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...